missing translation for 'onlineSavingsMsg'
Learn More

4-[(6-Methylpyrazin-2-yl)oxy]benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC62701DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC62701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

906353-00-2
230.22
PPGBPVGCYRDKMI-UHFFFAOYSA-N
24229686
CC1=CN=CC(OC2=CC=C(C=C2)C(O)=O)=N1
C12H10N2O3
MFCD09817529
4-6-methylpyrazin-2-yl oxy benzoic acid, 4-6-methylpyrazin-2-yloxy benzoic acid
4-[(6-methylpyrazin-2-yl)oxy]benzoic acid

Specifications

4-[(6-Methylpyrazin-2-yl)oxy]benzoic acid
96.5
97%
C12H10N2O3
MFCD09817529
PPGBPVGCYRDKMI-UHFFFAOYSA-N
4-[(6-methylpyrazin-2-yl)oxy]benzoic acid
24229686
97%
906353-00-2
100.0
Amber Glass Bottle
1g
4-6-methylpyrazin-2-yl oxy benzoic acid, 4-6-methylpyrazin-2-yloxy benzoic acid
CC1=CN=CC(OC2=CC=C(C=C2)C(O)=O)=N1
230.22
230.22
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents