missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-2-pyrone, 98%

Catalog No. AC295350100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295350100 10 g Glass bottle
AC295350010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295350100 Supplier Thermo Scientific Chemicals Supplier No. 295350100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 675-09-2 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 675-09-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00075555
Synonym 4,6-dimethyl-2-pyrone, 4,6-dimethyl-2h-pyran-2-one, mesitene lactone, 4,6-dimethylcoumalin, 2h-pyran-2-one, 4,6-dimethyl, 4,6-dimethyl-alpha-pyrone, 4,6-dimethyl-pyran-2-one, 2,4-dimethyl-alpha-pyrone, 2,4-dimethyl-.alpha.-pyrone, 4,6-dimethyl-.alpha.-pyrone

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 50°C
Color Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 124.14
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4, 6-Dimethyl-2-pyrone, 98%
Show More Show Less
EINECSNumber 211-618-7
RTECSNumber UQ0700000
TSCA TSCA
Recommended Storage Product may darken during storage

RUO – Research Use Only