missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dichloro-2-(trifluoromethyl)pyrimidine, 97%

Catalog No. AAH33142MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33142MD 250 mg
AAH3314203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33142MD Supplier Thermo Scientific Chemicals Supplier No. H33142MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 705-24-8 | C5HCl2F3N2 | 216.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 705-24-8
Molecular Formula C5HCl2F3N2
Molecular Weight (g/mol) 216.97
MDL Number MFCD08436597
InChI Key QFWVAJQVYBRTCL-UHFFFAOYSA-N
Synonym 4,6-dichloro-2-trifluoromethyl pyrimidine, 4,6-dichloro-2-trifluoromethyl-pyrimidine, pyrimidine, 4,6-dichloro-2-trifluoromethyl, pubchem19441, ksc496e6h, 4 pound not6-dichloro-2-trifluoromethylpyrimidine
PubChem CID 15713197
IUPAC Name 4,6-dichloro-2-(trifluoromethyl)pyrimidine
SMILES FC(F)(F)C1=NC(Cl)=CC(Cl)=N1

Specifications

Boiling Point 50°C to 52°C (5 mmHg)
Quantity 250 mg
Formula Weight 216.98
Percent Purity 97%
Chemical Name or Material 4,6-Dichloro-2-(trifluoromethyl)pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only