missing translation for 'onlineSavingsMsg'
Learn More

4,6-Diaminoresorcinol Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7126993
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18885G 5 g
D188825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18885G Supplier TCI America Supplier No. D18885G
Only null left

Chemical Identifiers

CAS 16523-31-2
Molecular Formula C6H10Cl2N2O2
Molecular Weight (g/mol) 213.058
MDL Number MFCD00143239
InChI Key KUMOYHHELWKOCB-UHFFFAOYSA-N
Synonym 4,6-diaminoresorcinol dihydrochloride, 4,6-diaminobenzene-1,3-diol dihydrochloride, 4,6-diaminoresorcinol 2hcl, 1,3-benzenediol, 4,6-diamino-, dihydrochloride, 4,6-diamino resorcinol dihydrochloride, 4,6-diamino-benzene-1,3-diol 2hcl salt, 1,3-benzenediol, 4,6-diamino-, hydrochloride 1:2, 4,6-bis azanyl benzene-1,3-diol dihydrochloride, pubchem20873, acmc-1c78d
PubChem CID 2733648
IUPAC Name 4,6-diaminobenzene-1,3-diol;dihydrochloride
SMILES C1=C(C(=CC(=C1N)O)O)N.Cl.Cl

Specifications

Quantity 5 g
Formula Weight 213.06
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Diaminoresorcinol Dihydrochloride
EINECSNumber (3)-4275
TSCA Yes