missing translation for 'onlineSavingsMsg'
Learn More

4(5)-(Trifluoromethyl)imidazole 98.0+%, TCI America™

Catalog No. T29281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29281G Supplier TCI America Supplier No. T29281G

Chemical Identifiers

CAS 33468-69-8
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.077
MDL Number MFCD08458860
InChI Key DFLGRTIPTPCKPJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1h-imidazole, 4-trifluoromethylimidazole, 4-trifluoromethyl imidazole, 5-trifluoromethyl-1h-imidazole, 4 5-trifluoromethyl imidazole, 1h-imidazole, 4-trifluoromethyl, imidazole, 4-trifluoromethyl, pubchem19080, acmc-1ailu
PubChem CID 585891
IUPAC Name 5-(trifluoromethyl)-1H-imidazole
SMILES C1=C(NC=N1)C(F)(F)F

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 1 g
Formula Weight 136.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4(5)-(Trifluoromethyl)imidazole