missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%

Catalog No. AAH33283MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33283MD 250 mg
AAH3328303 1 g
AAH3328306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33283MD Supplier Thermo Scientific Chemicals Supplier No. H33283MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 874838-96-7 | C12H7ClF3NO3S | 337.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874838-96-7
Molecular Formula C12H7ClF3NO3S
Molecular Weight (g/mol) 337.70
MDL Number MFCD07774162
InChI Key XEMAYRPWJCLKBQ-UHFFFAOYSA-N
Synonym 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-5-trifluoromethyl-2-pyridinyl-oxy benzenesulfonoyl chloride, 4-5-trifluoromethyl pyridin-2-yl oxy benzenesulphonyl chloride, trifluoromethylpyridinyloxybenzenesulfonoylchloride, 4-5-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, chloro 4-5-trifluoromethyl 2-pyridyloxy phenyl sulfone
PubChem CID 24213828
IUPAC Name 4-[5-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
SMILES FC(F)(F)C1=CN=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)C=C1

Specifications

Melting Point 51°C to 53°C
Quantity 250 mg
UN Number UN3261
Formula Weight 337.71
Percent Purity 97%
Chemical Name or Material 4-(5-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only