missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzylamine, 97%, Thermo Scientific™

Numéro de catalogue. p-3629701
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC61713CB 250mg Amber Glass Bottle
CC61713DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC61713CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC61713CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 946409-19-4
Formule moléculaire C10H11N3O
Poids moléculaire (g/mol) 189.218
Numéro MDL MFCD11109325
Clé InChI SBNUQIHHPTZLSV-UHFFFAOYSA-N
Synonyme 4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, 1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine, 4-5-methyl-1,3,4-oxadiazol-2-yl-benzenemethanamine
PubChem CID 33589501
Nom de l’IUPAC [4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
SOURIRES CC1=NN=C(O1)C2=CC=C(C=C2)CN

Spécifications

Nom ou substance chimique 4-(5-Methyl-1, 3, 4-oxadiazol-2-yl)benzylamine
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 189.22
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.