missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrophenylacetonitrile, 98%

CAS: 17354-04-0 | C10H10N2O4 | 222.20 g/mol

$68.14 - $178.30

Chemical Identifiers

CAS 17354-04-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00033967
InChI Key PKDJSFDIQCAPSY-UHFFFAOYSA-N
Synonym 2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
PubChem CID 222992
IUPAC Name 2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
SMILES COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH32747MD
View Documents
Thermo Scientific Chemicals
H32747MD
250 mg
Each for $68.14
Only null left
 
AAH3274703
View Documents
Thermo Scientific Chemicals
H3274703
1 g
Each for $178.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17354-04-0
222.20
PKDJSFDIQCAPSY-UHFFFAOYSA-N
222992
COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O
C10H10N2O4
MFCD00033967
2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile

Specifications

17354-04-0
C10H10N2O4
250 mg
2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O
222.20
222.2
4,5-Dimethoxy-2-nitrophenylacetonitrile
110°C to 112°C
MFCD00033967
UN3439
PKDJSFDIQCAPSY-UHFFFAOYSA-N
2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
222992
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only