missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrophenylacetonitrile, 98%

CAS: 17354-04-0 | C10H10N2O4 | 222.20 g/mol

Supplier:  Thermo Scientific Chemicals H3274703

 View more versions of this product

Catalog No. AAH3274703

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4,5-Dimethoxy-2-nitrophenylacetonitrile
110°C to 112°C
MFCD00033967
UN3439
PKDJSFDIQCAPSY-UHFFFAOYSA-N
2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
222992
98%
17354-04-0
C10H10N2O4
1 g
2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O
222.20
222.2

Chemical Identifiers

17354-04-0
222.20
PKDJSFDIQCAPSY-UHFFFAOYSA-N
222992
COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O
C10H10N2O4
MFCD00033967
2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only