missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethoxy-2-nitrophenylacetonitrile, 98%

Catalog No. AAH32747MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32747MD 250 mg
AAH3274703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32747MD Supplier Thermo Scientific Chemicals Supplier No. H32747MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17354-04-0 | C10H10N2O4 | 222.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17354-04-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00033967
InChI Key PKDJSFDIQCAPSY-UHFFFAOYSA-N
Synonym 2-4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenyl acetonitrile, 4,5-dimethoxy-2-nitrophenylacetonitrile, benzeneacetonitrile,4,5-dimethoxy-2-nitro, benzeneacetonitrile, 4,5-dimethoxy-2-nitro, acmc-1bvbg, 2-nitro-4,5-dimethoxybenzyl cyanide, 2-4,5-dimethoxy-2-nitrophenyl ethanenitrile, 4,5-dimethoxy-2-nitro-phenyl-acetonitrile
PubChem CID 222992
IUPAC Name 2-(4,5-dimethoxy-2-nitrophenyl)acetonitrile
SMILES COC1=CC(CC#N)=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 110°C to 112°C
Quantity 250 mg
UN Number UN3439
Formula Weight 222.2
Percent Purity 98%
Chemical Name or Material 4,5-Dimethoxy-2-nitrophenylacetonitrile
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only