missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dihydroxyanthraquinone-2-carboxylic Acid 95.0+%, TCI America™
$74.56 - $218.10
Chemical Identifiers
| CAS | 478-43-3 |
|---|---|
| Molecular Formula | C15H8O6 |
| Molecular Weight (g/mol) | 284.223 |
| MDL Number | MFCD00009618 |
| InChI Key | FCDLCPWAQCPTKC-UHFFFAOYSA-N |
| Synonym | rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid |
| PubChem CID | 10168 |
| ChEBI | CHEBI:8825 |
| IUPAC Name | 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid |
| SMILES | C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3986200MG
|
TCI America
D3986200MG |
200 mg |
Each for $74.56
|
|
|||||
|
D39861G
|
TCI America
D39861G |
1 g |
Each for $218.10
|
|
|||||
Chemical Identifiers
| 478-43-3 | |
| 284.223 | |
| FCDLCPWAQCPTKC-UHFFFAOYSA-N | |
| 10168 | |
| 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid |
| C15H8O6 | |
| MFCD00009618 | |
| rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid | |
| CHEBI:8825 | |
| C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O |
Specifications
| 478-43-3 | |
| C15H8O6 | |
| 200 mg | |
| FCDLCPWAQCPTKC-UHFFFAOYSA-N | |
| 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid | |
| 10168 | |
| 284.22 | |
| Crystalline Powder |
| 321°C | |
| MFCD00009618 | |
| rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid | |
| C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O | |
| 284.223 | |
| CHEBI:8825 | |
| ≥95.0% (HPLC,T) | |
| 4,5-Dihydroxyanthraquinone-2-carboxylic Acid |
Safety and Handling
RTECSNumber : CA9516000
TSCA : No