accessibility menu, dialog, popup

UserName

CAS RN 478-43-3

IUPAC Name: 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
Synonyms: monorhein cassic acid rhein (molecule) rheic acid rhein chrysazin-3-carboxylic acid rhubarb yellow dipropionyl rhein
Molecular Weight (g/mol): 284.22
Molecular Formula: C15H8O6
InChi Key: FCDLCPWAQCPTKC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C2C(=O)C3=C(O)C=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)