missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dihydroxyanthraquinone-2-carboxylic Acid 95.0+%, TCI America™
Supplier: TCI America D39861G
Specifications
| 4,5-Dihydroxyanthraquinone-2-carboxylic Acid | |
| 321°C | |
| MFCD00009618 | |
| rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid | |
| C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O | |
| 284.223 | |
| CHEBI:8825 | |
| ≥95.0% (HPLC,T) |
| 478-43-3 | |
| C15H8O6 | |
| 1 g | |
| FCDLCPWAQCPTKC-UHFFFAOYSA-N | |
| 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid | |
| 10168 | |
| 284.22 | |
| Crystalline Powder |
Chemical Identifiers
| 478-43-3 | |
| 284.223 | |
| FCDLCPWAQCPTKC-UHFFFAOYSA-N | |
| 10168 | |
| 4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid |
| C15H8O6 | |
| MFCD00009618 | |
| rhein, monorhein, rheic acid, rhubarb yellow, cassic acid, chrysazin-3-carboxylic acid, 4,5-dihydroxyanthraquinone-2-carboxylic acid, 4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, rheinic acid, 1,8-dihydroxyanthraquinone-3-carboxylic acid | |
| CHEBI:8825 | |
| C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O |
Safety and Handling
RTECSNumber : CA9516000
TSCA : No