missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dihydro-6-methyl-3(2H)-pyridazinone 98.0+%, TCI America™

Catalog No. D42165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42165G 5 g
D421625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42165G Supplier TCI America Supplier No. D42165G
Only null left

Chemical Identifiers

CAS 5157-08-4
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.13
MDL Number MFCD00006455
InChI Key VOTFXESXPPEARL-UHFFFAOYSA-N
Synonym 6-methyl-4,5-dihydropyridazin-3 2h-one, 4,5-dihydro-6-methylpyridazin-3 2h-one, 6-methyl-2,3,4,5-tetrahydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dihydro-6-methyl, 4,5-dihydro-6-methyl-3 2h-pyridazinone, 6-methyl-4,5-dihydro-2h-pyridazin-3-one, 6-methyl-4,5-dihydro-3 2h-pyridazinone, wln: t6nmv futj f1, 6-methyl-2,4,5-trihydropyridazin-3-one, 3 2h-pyridazinone,5-dihydro-6-methyl
PubChem CID 78834
IUPAC Name 3-methyl-4,5-dihydro-1H-pyridazin-6-one
SMILES CC1=NNC(=O)CC1

Specifications

Melting Point 81°C
Color White
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dihydro-6-methyl-3(2H)-pyridazinone
RTECSNumber UR6375000
TSCA No