missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichlorophthalic anhydride, 98%

Catalog No. p-3598200
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC387120050 5 g Glass bottle
AC387120250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC387120050 Supplier Thermo Scientific Chemicals Supplier No. 387120050

CAS: 942-06-3 | C8H2Cl2O3 | 217.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 942-06-3
Molecular Formula C8H2Cl2O3
Molecular Weight (g/mol) 217.00
MDL Number MFCD00075034
InChI Key ULSOWUBMELTORB-UHFFFAOYSA-N
Synonym 4,5-dichlorophthalic anhydride, 5,6-dichloroisobenzofuran-1,3-dione, 4,5-dichlorophthalicanhydride, 1,3-isobenzofurandione, 5,6-dichloro, 5,6-dichloro-1,3-dihydro-2-benzofuran-1,3-dione, 5,6-dichloro-1,3-dihydroisobenzofuran-1,3-dione, 5,6-dichlor-2-benzofuran-1,3-dion, pubchem16761, acmc-209rqk, 4,5-dichlorophalic anhydride
PubChem CID 70334
IUPAC Name 5,6-dichloro-2-benzofuran-1,3-dione
SMILES ClC1=CC2=C(C=C1Cl)C(=O)OC2=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 185°C to 187°C
Color Blue to Gray
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 217.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 5-Dichlorophthalic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-386-2

RUO – Research Use Only