missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-nitrotoluene 98.0+%, TCI America™

Catalog No. D32715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32715G 5 g
D327125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32715G Supplier TCI America Supplier No. D32715G
Only null left

Chemical Identifiers

CAS 7494-45-3
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.022
InChI Key AOTWCYSQDSKAQT-UHFFFAOYSA-N
PubChem CID 343795
IUPAC Name 1,2-dichloro-4-methyl-5-nitrobenzene
SMILES CC1=CC(=C(C=C1[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 62°C
Quantity 5 g
Formula Weight 206.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dichloro-2-nitrotoluene