missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-2-nitroaniline, 98%, Thermo Scientific Chemicals

Catalog No. AAA1790806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1790806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1790806 Supplier Thermo Scientific Chemicals Supplier No. A1790806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6641-64-1 | Molecular Formula: C6H4Cl2N2O2 | Molecular Weight: 207.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6641-64-1
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.01
MDL Number MFCD00007770
InChI Key FSGTULQLEVAYRS-UHFFFAOYSA-N
Synonym 3,4-dichloro-6-nitroaniline, benzenamine, 4,5-dichloro-2-nitro, unii-3i0g2adu1l, 2-nitro-4,5-dichloroaniline, 3i0g2adu1l, 4,5-dichloro-2-nitro-aniline, 4,5-dichloro-2-nitrobenzenamine, 4,5-dichlor-2-nitroanilin, pubchem3466, acmc-1b6pu
PubChem CID 81149
IUPAC Name 4,5-dichloro-2-nitroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)[N+](=O)[O-])N

Specifications

Melting Point 180°C to 182°C
Quantity 5 g
Beilstein 643127
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material 4,5-Dichloro-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 229-657-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only