missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dibromo-3(2H)-pyridazinone, 98%

Catalog No. AAA1599414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1599414 25 g
AAA1599406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1599414 Supplier Thermo Scientific Chemicals Supplier No. A1599414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5788-58-9 | C4H2Br2N2O | 253.881 g/mol

4,5-Dibromo-3(2H)-pyridazinone is used as an intermediate in the manufacture of pharmaceuticals. It is widely used in chemical research and in fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dibromo-3(2H)-pyridazinone is used as an intermediate in the manufacture of pharmaceuticals. It is widely used in chemical research and in fine chemicals.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5788-58-9
Molecular Formula C4H2Br2N2O
Molecular Weight (g/mol) 253.881
MDL Number MFCD00023641
InChI Key AGLQURQNVJVJNB-UHFFFAOYSA-N
Synonym 4,5-dibromopyridazin-3 2h-one, 4,5-dibromo-3 2h-pyridazinone, 4,5-dibromopyridazin-3-one, 4,5-dibromopyridazine-3 2h-one, 4,5-dibromopyridazin-3-ol, 4,5-dibromo-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dibromo, 4,5-dibromo-2h-pyridazin-3-one, 4,5-dibromo-2-hydropyridazin-3-one
PubChem CID 236181
IUPAC Name 4,5-dibromo-1H-pyridazin-6-one
SMILES C1=NNC(=O)C(=C1Br)Br

Specifications

Melting Point ∼220°C (decomposition)
Quantity 25 g
Beilstein 121469
Formula Weight 253.89
Percent Purity 98%
Chemical Name or Material 4,5-Dibromo-3(2H)-pyridazinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only