missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(mercaptomethyl)-o-xylene 96.0+%, TCI America™

Catalog No. B11051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B11051G Supplier TCI America Supplier No. B11051G
Only null left

Chemical Identifiers

CAS 10230-61-2
Molecular Formula C10H14S2
Molecular Weight (g/mol) 198.34
MDL Number MFCD00004869
InChI Key NNGQXIUDFKIVKI-UHFFFAOYSA-N
Synonym 1,2-Dimethyl-4,5-bis(mercaptomethyl)benzene, 4,5-Dimethyl-o-xylylenedithiol, Durene-alpha1,alpha2-dithiol
PubChem CID 82471
IUPAC Name [4,5-dimethyl-2-(sulfanylmethyl)phenyl]methanethiol
SMILES CC1=CC(CS)=C(CS)C=C1C

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 198.34
Percent Purity ≥96.0% (GC,T)
Chemical Name or Material 4,5-Bis(mercaptomethyl)-o-xylene