missing translation for 'onlineSavingsMsg'
Learn More

(+)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane 97.0+%, TCI America™

Catalog No. B20481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20481G 1 g
B20485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20481G Supplier TCI America Supplier No. B20481G

Chemical Identifiers

CAS 93379-49-8
Molecular Formula C31H30O4
Molecular Weight (g/mol) 466.577
MDL Number MFCD00010079
InChI Key OWVIRVJQDVCGQX-NSOVKSMOSA-N
PubChem CID 9852051
IUPAC Name [(4S,5S)-5-[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolan-4-yl]-diphenylmethanol
SMILES CC1(OC(C(O1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O)C

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 1 g
Synonym (+)-trans-alpha,alpha′C-(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(diphenylmethanol), (2S,3S)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-1,2,3,4-butanetetrol, (+)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-D-threitol
Formula Weight 466.58
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane