missing translation for 'onlineSavingsMsg'
Learn More

(-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane 97.0+%, TCI America™

Catalog No. B16141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16141G 1 g
B16145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16141G Supplier TCI America Supplier No. B16141G

Chemical Identifiers

CAS 93379-48-7
Molecular Formula C31H30O4
Molecular Weight (g/mol) 466.577
MDL Number MFCD00064467
InChI Key OWVIRVJQDVCGQX-VSGBNLITSA-N
PubChem CID 2725026
IUPAC Name [(4R,5R)-5-[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolan-4-yl]-diphenylmethanol
SMILES CC1(OC(C(O1)C(C2=CC=CC=C2)(C3=CC=CC=C3)O)C(C4=CC=CC=C4)(C5=CC=CC=C5)O)C

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 1 g
Synonym (-)-trans-alpha,alpha′C-(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(diphenylmethanol), (2R,3R)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-1,2,3,4-butanetetrol, (-)-2,3-O-Isopropylidene-1,1,4,4-tetraphenyl-L-threitol, TADDOL
Formula Weight 466.58
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-4,5-Bis[hydroxy(diphenyl)methyl]-2,2-dimethyl-1,3-dioxolane