missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Oxydiphthalic Anhydride (purified by sublimation) 99.0+%, TCI America™

Catalog No. O04235G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04235G Supplier TCI America Supplier No. O04235G
Only null left

Chemical Identifiers

CAS 1823-59-2
Molecular Formula C16H6O7
Molecular Weight (g/mol) 310.217
MDL Number MFCD00039144
InChI Key QQGYZOYWNCKGEK-UHFFFAOYSA-N
PubChem CID 74574
IUPAC Name 5-[(1,3-dioxo-2-benzofuran-5-yl)oxy]-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1OC3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point 229°C
Color White
Quantity 5 g
Formula Weight 310.22
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Oxydiphthalic Anhydride (purified by sublimation)
EINECSNumber (3)-3937
TSCA Yes