missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Oxybis(benzoic acid), 98+%

Catalog No. AAL1565122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1565122 100 g
AAL1565114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1565122 Supplier Thermo Scientific Chemicals Supplier No. L1565122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2215-89-6 | C14H10O5 | 258.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2215-89-6
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.229
MDL Number MFCD00013988
InChI Key WVDRSXGPQWNUBN-UHFFFAOYSA-N
Synonym 4,4'-oxybis benzoic acid, 4,4'-oxydibenzoic acid, 4,4'-oxybisbenzoic acid, 4-4-carboxyphenoxy benzoic acid, benzoic acid, 4,4'-oxybis, 4,4'-dicarboxydiphenyl ether, 4,4-oxobisbenzoic acid, 4,4'-diphenyl ether dicarboxylic acid, diphenyl ether 4,4'-dicarboxylic acid, pubchem9032
PubChem CID 75183
IUPAC Name 4-(4-carboxyphenoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 329°C to 332°C
Quantity 100 g
Beilstein 2218315
Formula Weight 258.23
Percent Purity ≥98%
Chemical Name or Material 4,4'-Oxybis(benzoic acid)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-683-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only