missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazinyl)benzaldehyde 98.0+%, TCI America™

Numéro de catalogue. M16245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M16245G 5 g
M162425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M16245G Fournisseur TCI America Code fournisseur M16245G
Il en reste null

Identifiants chimiques

CAS 27913-99-1
Molecular Formula C12H17N2O
Molecular Weight (g/mol) 205.28
MDL Number MFCD00452233
InChI Key PFODEVGLOVUVHS-UHFFFAOYSA-O
Synonym 4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr
PubChem CID 736533
IUPAC Name 4-(4-formylphenyl)-1-methylpiperazin-1-ium
SMILES C[NH+]1CCN(CC1)C1=CC=C(C=O)C=C1

Spécifications

Melting Point 63°C
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Methylpiperazinyl)benzaldehyde