missing translation for 'onlineSavingsMsg'
Learn More

{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™

Numéro de catalogue. p-3522392
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC55809DA 1g Amber Glass Bottle
CC55809DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC55809DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC55809DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 884507-50-0
Formule moléculaire C14H22N2O
Poids moléculaire (g/mol) 234.343
Numéro MDL MFCD09064973
Clé InChI YQUWAYPNHCWYNU-UHFFFAOYSA-N
Synonyme 4-4-methylperhydro-1,4-diazepin-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazepan-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazaperhydroepinyl methyl phenyl methan-1-ol, benzenemethanol,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl
PubChem CID 24229552
Nom de l’IUPAC [4-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl]methanol
SOURIRES CN1CCCN(CC1)CC2=CC=C(C=C2)CO

Spécifications

Nom ou substance chimique {4-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]phenyl}methanol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 234.34
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.