missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)benzoic Acid 98.0+%, TCI America™

Catalog No. H06955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06955G 5 g
H069525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06955G Supplier TCI America Supplier No. H06955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58574-03-1
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00059078
InChI Key JTGCXYYDAVPSFD-UHFFFAOYSA-N
Synonym 4'-hydroxy-4-biphenylcarboxylic acid, 4-4-hydroxyphenyl benzoic acid, 4'-hydroxybiphenyl-4-carboxylic acid, 4'-hydroxy-1,1'-biphenyl-4-carboxylic acid, 4-hydroxy-4'-biphenylcarboxylic acid, 4'-hydroxy biphenyl-4-carboxylic acid, 4-hydroxybiphenyl-4'-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-hydroxy, chembl90522, 4'-hydroxy 1,1'-biphenyl-4-carboxylic acid
PubChem CID 301556
IUPAC Name 4-(4-hydroxyphenyl)benzoic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 214.22
Percent Purity ≥98.0% (T,GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxyphenyl)benzoic Acid