missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)-2-butanone 99.0+%, TCI America™

Supplier:  TCI America H060425G

 View more versions of this product

Catalog No. H060425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(4-Hydroxyphenyl)-2-butanone
83°C
C10H12O2
25 g
NJGBTKGETPDVIK-UHFFFAOYSA-N
4-(4-hydroxyphenyl)butan-2-one
21648
164.20
Crystalline Powder
5471-51-2
White
MFCD00002394
4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
CC(=O)CCC1=CC=C(C=C1)O
164.204
CHEBI:68656
≥99.0% (GC)

Chemical Identifiers

5471-51-2
164.204
NJGBTKGETPDVIK-UHFFFAOYSA-N
21648
4-(4-hydroxyphenyl)butan-2-one
C10H12O2
MFCD00002394
4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
CHEBI:68656
CC(=O)CCC1=CC=C(C=C1)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2930

RTECSNumber : EL8925000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions