missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)-2-butanone 99.0+%, TCI America™

Catalog No. p-7130631
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H060425G 25 g
H0604250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H060425G Supplier TCI America Supplier No. H060425G
Only null left

Chemical Identifiers

CAS 5471-51-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002394
InChI Key NJGBTKGETPDVIK-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
PubChem CID 21648
ChEBI CHEBI:68656
IUPAC Name 4-(4-hydroxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 83°C
Color White
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-butanone
EINECSNumber (3)-2930
RTECSNumber EL8925000
TSCA Yes