missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Hydroxyphenyl)-2-butanone 99.0+%, TCI America™
Supplier: TCI America H0604250G
Specifications
| 4-(4-Hydroxyphenyl)-2-butanone | |
| 83°C | |
| C10H12O2 | |
| 250 g | |
| NJGBTKGETPDVIK-UHFFFAOYSA-N | |
| 4-(4-hydroxyphenyl)butan-2-one | |
| 21648 | |
| 164.20 | |
| Crystalline Powder |
| 5471-51-2 | |
| White | |
| MFCD00002394 | |
| 4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone | |
| CC(=O)CCC1=CC=C(C=C1)O | |
| 164.204 | |
| CHEBI:68656 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 5471-51-2 | |
| 164.204 | |
| NJGBTKGETPDVIK-UHFFFAOYSA-N | |
| 21648 | |
| 4-(4-hydroxyphenyl)butan-2-one |
| C10H12O2 | |
| MFCD00002394 | |
| 4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone | |
| CHEBI:68656 | |
| CC(=O)CCC1=CC=C(C=C1)O |
Safety and Handling
EINECSNumber : (3)-2930
RTECSNumber : EL8925000
TSCA : Yes