missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Hexamethylenebis(1,1-dimethylsemicarbazide) 98.0+%, TCI America™

Catalog No. H1064500G
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
H1064500G 500 g
1 options

Catalog No. H1064500G

Supplier: TCI America H1064500G

Only null left

Chemical Identifiers

CAS 69938-76-7
Molecular Formula C12H28N6O2
Molecular Weight (g/mol) 288.40
MDL Number MFCD00089122
InChI Key VETHREXFBVHLJJ-UHFFFAOYSA-N
Synonym 1,6-Bis[3-(dimethylamino)ureido]hexane
PubChem CID 4404942
IUPAC Name 3-(dimethylamino)-1-{6-[(N',N'-dimethylhydrazinecarbonyl)amino]hexyl}urea
SMILES CN(C)NC(=O)NCCCCCCNC(=O)NN(C)C

Specifications

Melting Point 146°C
Color White
Quantity 500 g
Formula Weight 288.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Hexamethylenebis(1,1-dimethylsemicarbazide)
EINECSNumber (2)-1771
TSCA Yes