missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Hexafluoroisopropylidene)diphenol, 98%

Catalog No. AAA1837022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1837022 100 g
AAA1837006 5 g
AAA1837014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1837022 Supplier Thermo Scientific Chemicals Supplier No. A1837022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1478-61-1 | C15H10F6O2 | 336.23 g/mol

4,4'-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1478-61-1
Molecular Formula C15H10F6O2
Molecular Weight (g/mol) 336.23
MDL Number MFCD00000439
InChI Key ZFVMWEVVKGLCIJ-UHFFFAOYSA-N
Synonym bisphenol af, 4,4'-hexafluoroisopropylidene diphenol, 2,2-bis 4-hydroxyphenyl hexafluoropropane, hexafluorobisphenol a, biphenol af, 4,4'-perfluoropropane-2,2-diyl diphenol, hexafluoroisopropylidenebis 4-hydroxybenzene, hexafluorodiphenylolpropane, hexafluoroacetone bisphenol a, 2,2-bis 4-hydroxyphenyl perfluoropropane
PubChem CID 73864
ChEBI CHEBI:72754
IUPAC Name 4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 158°C to 162°C
Boiling Point 400°C
Quantity 100 g
Beilstein 1891568
Solubility Information Insoluble in water.
Formula Weight 336.24
Percent Purity 98%
Chemical Name or Material 4,4'-(Hexafluoroisopropylidene)diphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-036-7
RTECSNumber SN2780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only