missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[[(4-Fluorophenyl)imino]methyl]phenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 181°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 215.23 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-[[(4-Fluorophenyl)imino]methyl]phenol |
Safety and Handling
| EINECSNumber | (3)-4534 |
| TSCA | No |