missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dithiodibutyric Acid 95.0+%, TCI America™

Catalog No. D17575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17575G 5 g
D175725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17575G Supplier TCI America Supplier No. D17575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2906-60-7
Molecular Formula C8H14O4S2
Molecular Weight (g/mol) 238.316
MDL Number MFCD00004406
InChI Key YYSCJLLOWOUSHH-UHFFFAOYSA-N
PubChem CID 76196
IUPAC Name 4-(3-carboxypropyldisulfanyl)butanoic acid
SMILES C(CC(=O)O)CSSCCCC(=O)O

Specifications

Melting Point 108°C
Quantity 5 g
Synonym 4,4'-dithiodibutyric acid, butanoic acid, 4,4'-dithiobis, 4,4'-disulfanediyldibutanoic acid, 3-carboxypropyl disulfide, 4-3-carboxypropyl disulfanyl butanoic acid, 4,4-dithiodibutyric acid, butanoic acid,4,4'-dithiobis, bis 3-carboxypropyl disulfide, 4,4'-dithiodibutanoic acid, butyric acid, 4,4'-dithiodi
Formula Weight 238.32
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dithiodibutyric Acid