missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dipyridyl Disulfide 97.0+%, TCI America™

Catalog No. D24775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24775G 5 g
D247725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24775G Supplier TCI America Supplier No. D24775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2645-22-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.308
MDL Number MFCD00006423
InChI Key UHBAPGWWRFVTFS-UHFFFAOYSA-N
Synonym 4,4'-dithiodipyridine, 4,4'-dipyridyl disulfide, 1,2-di pyridin-4-yl disulfane, aldrithiol-4, pyridine, 4,4'-dithiobis, 4,4'-dithiopyridine, di 4-pyridyl disulfide, aldrithiol 4, di 4-pyridinyl disulfide, 4,4'-dipyridine disulfide
PubChem CID 75846
ChEBI CHEBI:41814
IUPAC Name 4-(pyridin-4-yldisulfanyl)pyridine
SMILES C1=CN=CC=C1SSC2=CC=NC=C2

Specifications

Melting Point 75°C
Quantity 5 g
Formula Weight 220.31
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dipyridyl Disulfide
TSCA No
Recommended Storage Refrigerator