missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dinitrostilbene 98.0+%, TCI America™

Numéro de catalogue. D24645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D24645G 5 g
D246425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D24645G Fournisseur TCI America Code fournisseur D24645G
Il en reste null

Identifiants chimiques

CAS 2501-02-2
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.24
MDL Number MFCD00017043
InChI Key CLVIIRIMEIEKOQ-OWOJBTEDSA-N
PubChem CID 5377860
ChEBI CHEBI:79798
IUPAC Name 1-nitro-4-[(E)-2-(4-nitrophenyl)ethenyl]benzene
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Spécifications

Melting Point 295°C
Color Yellow
Quantity 5 g
Formula Weight 270.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinitrostilbene