missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzophenone 99.0+%, TCI America™

Catalog No. D06855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06855G 5 g
D068525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06855G Supplier TCI America Supplier No. D06855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-97-2
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017214
InChI Key ZWPWLKXZYNXATK-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzophenone, p-tolyl ketone, bis 4-methylphenyl methanone, di-p-tolyl ketone, methanone, bis 4-methylphenyl, p,p'-dimethylbenzophenone, p,p'-dimethyl di-phenyl ketone, bis-p-tolylmethanone, benzophenone, 4,4'-dimethyl, 4,4-dimethylbenzophenone
PubChem CID 69148
IUPAC Name bis(4-methylphenyl)methanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 151°C
Quantity 5 g
Formula Weight 210.28
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbenzophenone