missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-2-cyclohexen-1-one 96.0+%, TCI America™

Catalog No. D38525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38525G 5 g
D385225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38525G Supplier TCI America Supplier No. D38525G

Chemical Identifiers

CAS 1073-13-8
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.18
MDL Number MFCD00009695
InChI Key HAUNPYVLVAIUOO-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-cyclohexen-1-one, 4,4-dimethylcyclohex-2-enone, 2-cyclohexen-1-one, 4,4-dimethyl, 4,4-dimethyl-2-cyclohexenone, 4,4-dimethylcyclohexenone, 4,4-dimethyl-2-cyclohexene-1-one, 4,4-dimethyl-cyclohex-2-en-1-one, pubchem17152, acmc-1bxzo, 3,3-dimethylcyclohexene-6-one
PubChem CID 136839
IUPAC Name 4,4-dimethylcyclohex-2-en-1-one
SMILES CC1(C)CCC(=O)C=C1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 124.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-2-cyclohexen-1-one