missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethyl-1-pentene 99.0+%, TCI America™

Catalog No. D12525ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12525ML 5 mL
D12521ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12525ML Supplier TCI America Supplier No. D12525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 762-62-9
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00026337
InChI Key KLCNJIQZXOQYTE-UHFFFAOYSA-N
Synonym 4,4-dimethyl-1-pentene, 1-pentene, 4,4-dimethyl, 2,2-dimethyl-4-pentene, 4,4-dimethyl-pent-1-ene, 4,4-dimethyl pent-1-ene, neoheptaene, acmc-1bcwv, ch3 3cch2ch=ch2, 4,4'-dimethyl-1-pentene
PubChem CID 12984
IUPAC Name 4,4-dimethylpent-1-ene
SMILES CC(C)(C)CC=C

Specifications

Color Colorless
Boiling Point 72°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-1-pentene