missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxychalcone, 99%

Catalog No. AAL1058518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1058518 50 g
AAL1058509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1058518 Supplier Thermo Scientific Chemicals Supplier No. L1058518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2373-89-9 | C17H16O3 | 268.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2373-89-9
Molecular Formula C17H16O3
Molecular Weight (g/mol) 268.31
MDL Number MFCD00025815
InChI Key HDXVSZWKIHQDES-LFYBBSHMSA-N
Synonym 4,4'-dimethoxychalcone, e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 2e-1,3-bis 4-methoxyphenyl prop-2-en-1-one, 1,3-bis 4-methoxyphenyl prop-2-en-1-one, 4,4/'-dimethoxychalcone, 2-propen-1-one, 1,3-bis 4-methoxyphenyl, e-4,4'-dimethoxychalcone, 1,3-bis 4-methoxyphenyl-2-propen-1-one, e-1,3-bis 4-methoxyphenyl-2-propen-1-one, 2e-1,3-bis 4-methoxyphenyl-2-propen-1-one
PubChem CID 5377817
IUPAC Name (E)-1,3-bis(4-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=C(\C=C\C(=O)C2=CC=C(OC)C=C2)C=C1

Specifications

Melting Point 99°C to 103°C
Quantity 50 g
Beilstein 1622293
Formula Weight 268.32
Percent Purity 99%
Chemical Name or Material 4,4'-Dimethoxychalcone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only