missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzophenone, 97%

Catalog No. AC161740250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161740250 25 g Plastic bottle
AC161741000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161740250 Supplier Thermo Scientific Chemicals Supplier No. 161740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 90-96-0 | C15H14O3 | 242.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-96-0
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00008404
InChI Key RFVHVYKVRGKLNK-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzophenone, bis 4-methoxyphenyl methanone, methanone, bis 4-methoxyphenyl, p,p'-dimethoxybenzophenone, dmbp, bis p-anisyl ketone, bis 4-anisyl ketone, bis p-methoxy benzophenone, benzophenone, 4,4'-dimethoxy, 1,1-di p-methoxyphenyl-ketone
PubChem CID 7032
IUPAC Name bis(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 141.0°C to 146.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula (CH3OC6H4)2CO
Quantity 25 g
Beilstein 08, 317
Formula Weight 242.27
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4,4′-Dimethoxybenzophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-028-0
TSCA TSCA

RUO – Research Use Only