Learn More
4,4'-Dihydroxybenzophenone, 98+%
CAS: 611-99-4 | C13H10O3 | 214.22 g/mol
Supplier: Thermo Scientific Chemicals A1509714
Description
4,4'-Dihydroxybenzophenone is as a UV light stabilizer. It and its derivatives are found in cosmetics, plastics, films, adhesives and coatings, optical fiber, and printed circuit boards. It is the precursor to certain polycarbonate polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-Dihydroxybenzophenone is as a UV light stabilizer. It and its derivatives are found in cosmetics, plastics, films, adhesives and coatings, optical fiber, and printed circuit boards. It is the precursor to certain polycarbonate polymers.
Solubility
Soluble in water(0.45g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 4,4'-Dihydroxybenzophenone | |
| 221°C to 225°C | |
| MFCD00002358 | |
| 1874572 | |
| Soluble in water(0.45g/L). | |
| C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O | |
| 214.22 | |
| CHEBI:34365 | |
| ≥98% |
| 611-99-4 | |
| C13H10O3 | |
| 25 g | |
| 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy | |
| RXNYJUSEXLAVNQ-UHFFFAOYSA-N | |
| bis(4-hydroxyphenyl)methanone | |
| 69150 | |
| 214.22 |
Chemical Identifiers
| 611-99-4 | |
| 214.22 | |
| RXNYJUSEXLAVNQ-UHFFFAOYSA-N | |
| 69150 | |
| bis(4-hydroxyphenyl)methanone |
| C13H10O3 | |
| MFCD00002358 | |
| 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy | |
| CHEBI:34365 | |
| C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-288-1
RTECSNumber : DJ0880000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only