accessibility menu, dialog, popup

UserName

CAS RN 611-99-4

IUPAC Name: 4-(4-hydroxybenzoyl)phenol
Synonyms: bis(4-hydroxyphenyl)methanone hbp (ketone) 4,4'-dihydroxydiphenyl ketone bis(p-hydroxy)benzophenone
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: RXNYJUSEXLAVNQ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)