missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxybenzophenone, 98+%

Catalog No. p-7022362
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1509706 5 g
AAA1509714 25 g
AAA1509722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1509706 Supplier Thermo Scientific Chemicals Supplier No. A1509706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-99-4 | C13H10O3 | 214.22 g/mol

4,4'-Dihydroxybenzophenone is as a UV light stabilizer. It and its derivatives are found in cosmetics, plastics, films, adhesives and coatings, optical fiber, and printed circuit boards. It is the precursor to certain polycarbonate polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dihydroxybenzophenone is as a UV light stabilizer. It and its derivatives are found in cosmetics, plastics, films, adhesives and coatings, optical fiber, and printed circuit boards. It is the precursor to certain polycarbonate polymers.

Solubility
Soluble in water(0.45g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 611-99-4
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002358
InChI Key RXNYJUSEXLAVNQ-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy
PubChem CID 69150
ChEBI CHEBI:34365
IUPAC Name bis(4-hydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O

Specifications

Melting Point 221°C to 225°C
Quantity 5 g
Beilstein 1874572
Solubility Information Soluble in water(0.45g/L).
Formula Weight 214.22
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dihydroxybenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-288-1
RTECSNumber DJ0880000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only