missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxybenzophénone, 97%

Numéro de catalogue. AC114920050
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
Packaging:
Bouteille en verre
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC114920050 5 g Bouteille en verre
AC114920250 25 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC114920050 Fournisseur Thermo Scientific Chemicals Code fournisseur 114920050

CAS : 611-99-4 | C13H10O 3 | 214,22 g/mol

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Identifiants chimiques

CAS 611-99-4
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002358
InChI Key RXNYJUSEXLAVNQ-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy
PubChem CID 69150
ChEBI CHEBI:34365
IUPAC Name Bis(4-hydroxyphényl)méthanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O

Spécifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 217°C to 222°C
Color Du blanc au beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Bouteille en verre
Linear Formula (HOC6H4)2CO
Quantity 5 g
Beilstein 08, 316
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in ether and ethanol
Formula Weight 214.22
Percent Purity 97%
Physical Form Poudre fine cristalline
Chemical Name or Material 4,4′-Dihydroxybenzophenone, 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-288-1
RTECSNumber DJ0880000
TSCA TSCA

Utilisation uniquement de la recherche RUO –