missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobenzophenone, 99%

CAS: 90-98-2 | C13H8Cl2O | 251.11 g/mol

Supplier:  Thermo Scientific Chemicals A1260836

 View more versions of this product

Catalog No. AAA1260836


Only null left
Explore available promotions

Description

Description

It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Soluble in hot acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Avoid strong oxidizing agents.
Specifications

Specifications

4,4'-Dichlorobenzophenone
144°C to 146°C
C13H8Cl2O
500 g
4,4'-dichlorobenzophenone, bis 4-chlorophenyl methanone, methanone, bis 4-chlorophenyl, bis 4-chlorophenyl ketone, dcbp, di p-chlorophenyl ketone, di 4-chlorophenyl ketone, bis p-chlorophenyl ketone, p,p'-dichlorobenzophenone, usaf do-4
OKISUZLXOYGIFP-UHFFFAOYSA-N
bis(4-chlorophenyl)methanone
7034
251.11
90-98-2
352°C to 354°C
MFCD00000623
643345
Soluble in hot acetone.
ClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
251.11
CHEBI:27519
99%

Chemical Identifiers

90-98-2
251.11
OKISUZLXOYGIFP-UHFFFAOYSA-N
7034
bis(4-chlorophenyl)methanone
C13H8Cl2O
MFCD00000623
4,4'-dichlorobenzophenone, bis 4-chlorophenyl methanone, methanone, bis 4-chlorophenyl, bis 4-chlorophenyl ketone, dcbp, di p-chlorophenyl ketone, di 4-chlorophenyl ketone, bis p-chlorophenyl ketone, p,p'-dichlorobenzophenone, usaf do-4
CHEBI:27519
ClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-030-1

RTECSNumber : DJ0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only