missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobenzophenone, 99%

Catalog No. p-7020597
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1260814 25 g
AAA1260822 100 g
AAA1260836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1260814 Supplier Thermo Scientific Chemicals Supplier No. A1260814

CAS: 90-98-2 | C13H8Cl2O | 251.11 g/mol

It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Soluble in hot acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Avoid strong oxidizing agents.

Chemical Identifiers

CAS 90-98-2
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.11
MDL Number MFCD00000623
InChI Key OKISUZLXOYGIFP-UHFFFAOYSA-N
Synonym 4,4'-dichlorobenzophenone, bis 4-chlorophenyl methanone, methanone, bis 4-chlorophenyl, bis 4-chlorophenyl ketone, dcbp, di p-chlorophenyl ketone, di 4-chlorophenyl ketone, bis p-chlorophenyl ketone, p,p'-dichlorobenzophenone, usaf do-4
PubChem CID 7034
ChEBI CHEBI:27519
IUPAC Name bis(4-chlorophenyl)methanone
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 144°C to 146°C
Boiling Point 352°C to 354°C
Quantity 25 g
Beilstein 643345
Solubility Information Soluble in hot acetone.
Formula Weight 251.11
Percent Purity 99%
Chemical Name or Material 4,4'-Dichlorobenzophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-030-1
RTECSNumber DJ0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only