accessibility menu, dialog, popup

UserName

ClOCl

CAS RN 90-98-2

IUPAC Name: bis(4-chlorophenyl)methanone
Synonyms: 4,4'-dichlorobenzophenone dbp (degradation product) C13H8cl2O usaf DO-4 DCBP
Molecular Weight (g/mol): 251.11
Molecular Formula: C13H8Cl2O
InChi Key: OKISUZLXOYGIFP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)