missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dicarboxydiphenyl Sulfone 98.0+%, TCI America™

Catalog No. D23675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23675G Supplier TCI America Supplier No. D23675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2449-35-6
Molecular Formula C14H10O6S
Molecular Weight (g/mol) 306.288
MDL Number MFCD00020375
InChI Key SQJQLYOMPSJVQS-UHFFFAOYSA-N
PubChem CID 224167
IUPAC Name 4-(4-carboxyphenyl)sulfonylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 370°C
Color White
Quantity 5 g
Synonym 4,4′C-Sulfonyldibenzoic Acid
Formula Weight 306.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dicarboxydiphenyl Sulfone