missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromooctafluorobiphenyl 95.0+%, TCI America™

Catalog No. p-7126883
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16961G 1 g
D16965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16961G Supplier TCI America Supplier No. D16961G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10386-84-2
Molecular Formula C12Br2F8
Molecular Weight (g/mol) 455.93
MDL Number MFCD00000310
InChI Key YXLMNFVUNLCJJY-UHFFFAOYSA-N
PubChem CID 82600
IUPAC Name 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl
SMILES FC1=C(F)C(=C(F)C(F)=C1Br)C1=C(F)C(F)=C(Br)C(F)=C1F

Specifications

Melting Point 115°C
Quantity 1 g
UN Number 3152
Synonym 4,4'-dibromooctafluorobiphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluorobiphenyl, octafluoro-4,4'-dibromobiphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro-1,1'-biphenyl, 1,1'-biphenyl, 4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro, 1,1'-biphenyl,4,4'-dibromo-2,2',3,3',5,5',6,6'-octafluoro, 1-bromo-4-4-bromo-2,3,5,6-tetrafluorophenyl-2,3,5,6-tetrafluorobenzene, f-pbb, 4,4/'-dibromooctafluorobiphenyl, 4,4\\'-dibromooctafluorobiphenyl
Formula Weight 455.93
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromooctafluorobiphenyl