missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Dibromo-1,1':3',1″-terphenyl 98.0+%, TCI America™

Catalog No. D5023200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5023200MG 200 mg
D50231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5023200MG Supplier TCI America Supplier No. D5023200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 83909-22-2
Molecular Formula C18H12Br2
Molecular Weight (g/mol) 388.102
MDL Number MFCD28138091
InChI Key XYRNOZKOAZFOOG-UHFFFAOYSA-N
Synonym 4,4′′-Dibromo-m-terphenyl
PubChem CID 10667976
IUPAC Name 1,3-bis(4-bromophenyl)benzene
SMILES C1=CC(=CC(=C1)C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br

Specifications

Melting Point 158°C
Color White
Quantity 200 mg
UN Number 3152
Formula Weight 388.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′′-Dibromo-1,1′:3′,1′′-terphenyl